Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 2 | 399 | ||
NM_001172767.2 | NP_001166238.1 | 1 | 35 | |||
NM_014491.3 | NP_055306.1 | 2 | 399 | |||
NM_148898.3 | NP_683696.2 | 2 | 399 | |||
NM_148899.3 | NP_683697.2 | 1 | 35 | |||
NM_148900.3 | NP_683698.2 | 2 | 399 | |||
NR_033766.1 | 3 | 428 | ||||
NR_033767.1 | 2 | 399 | ||||
XM_017012801.1 | XP_016868290.1 | 4 | 1005 | |||
GenBank mRNA | AF337817.1 | - | 2 | 175 | ||
AF454830.1 | - | 4 | 444 | |||
AF467252.1 | - | 2 | 126 | |||
AF467253.1 | - | 1 | 37 | |||
AF467257.1 | - | 2 | 77 | |||
AF467259.1 | - | 1 | 22 | |||
AF493430.1 | - | 2 | 192 | |||
AK131266.1 | - | 3 | 412 | |||
AK296957.1 | - | 2 | 279 | |||
AY144615.1 | - | 2 | 394 | |||
BC126104.1 | - | 2 | 73 | 8991820 | ||
BC143866.1 | - | 2 | 73 | 9052382 | ||
BC143867.1 | - | 2 | 73 | 9052383 | ||
CR749236.1 | - | 6 | 519 | |||
DA400243.1 | - | 3 | 428 | |||
KJ534839.1 | - | 1 | 25 | |||
KJ535079.1 | - | 1 | 25 | |||
KJ535086.1 | - | 1 | 24 |