Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127487.1 | NP_001120959.1 | 6 | 1218 | ||
NM_001458.4 | NP_001449.3 | 6 | 1218 | |||
GenBank mRNA | AB371585.1 | - | 6 | 1238 | ||
AF089841.1 | - | 6 | 1058 | |||
AJ012737.1 | - | 6 | 1027 |