Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127487.1 | NP_001120959.1 | 47 | 8187 | ||
NM_001458.4 | NP_001449.3 | 48 | 8286 | |||
GenBank mRNA | AB208865.1 | - | 25 | 3928 | ||
AB371585.1 | - | 48 | 8306 | |||
AF089841.1 | - | 48 | 8126 | |||
AF146692.1 | - | 47 | 8187 | |||
AJ012737.1 | - | 47 | 7996 | |||
AK001048.1 | - | 4 | 596 | |||
AK126540.1 | - | 1 | 2637 |