Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020661.2 | NP_065712.1 | 2 | 113 | ||
XM_011520772.2 | XP_011519074.1 | 2 | 232 | |||
XM_011520773.2 | XP_011519075.1 | 2 | 237 | |||
GenBank mRNA | AB040431.1 | - | 2 | 110 | ||
AF529815.1 | - | 1 | 34 | |||
AF529816.1 | - | 1 | 34 | |||
AF529817.1 | - | 1 | 34 | |||
AF529818.1 | - | 1 | 34 | |||
AF529819.1 | - | 1 | 34 | |||
AF529820.1 | - | 1 | 34 | |||
AF529821.1 | - | 1 | 34 | |||
AF529822.1 | - | 1 | 34 | |||
AF529823.1 | - | 1 | 34 | |||
AF529824.1 | - | 1 | 34 | |||
AF529825.1 | - | 1 | 34 | |||
AF529826.1 | - | 1 | 34 | |||
AF529827.1 | - | 1 | 34 | |||
AY534975.1 | - | 1 | 34 | |||
AY536516.1 | - | 1 | 34 | |||
AY536517.1 | - | 1 | 34 | |||
AY541058.1 | - | 1 | 34 | |||
BC006296.2 | - | 2 | 110 | 4054915 | ||
BT007402.1 | - | 1 | 34 | |||
BX402063.2 | - | 2 | 127 | |||
DQ431660.1 | - | 1 | 28 | |||
EU794587.1 | - | 2 | 110 |