Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020661.2 | NP_065712.1 | 4 | 549 | ||
XM_011520772.2 | XP_011519074.1 | 4 | 638 | |||
GenBank mRNA | AJ577811.1 | - | 2 | 262 |