Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000166.5 | NP_000157.1 | 2 | 225 | ||
NM_001097642.2 | NP_001091111.1 | 2 | 174 | |||
XM_011530907.2 | XP_011529209.1 | 2 | 273 | |||
XM_017029408.1 | XP_016884897.1 | 3 | 394 | |||
GenBank mRNA | AK313474.1 | - | 2 | 169 | ||
BC002805.1 | - | 2 | 134 | 3638623 | ||
BC022426.1 | - | 2 | 205 | 4252281 | ||
BC039198.1 | - | 2 | 147 | 4710239 | ||
BI764356.1 | - | 2 | 174 | |||
BT019329.1 | - | 1 | 79 | |||
DA081668.1 | - | 2 | 225 | |||
DA262106.1 | - | 2 | 225 | |||
X04325.1 | - | 2 | 141 |