Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008404.2 | NP_001008404.1 | 5 | 1255 | ||
NM_001286399.1 | NP_001273328.1 | 5 | 1211 | |||
NM_001286400.1 | NP_001273329.1 | 5 | 1420 | |||
XM_005267638.3 | XP_005267695.1 | 5 | 1324 | |||
XM_011536764.2 | XP_011535066.1 | 5 | 1302 | |||
GenBank mRNA | AK095913.1 | - | 5 | 1220 | ||
AK297818.1 | - | 5 | 1122 | |||
BC041103.1 | - | 5 | 1157 | 5193008 | ||
BX161457.1 | - | 5 | 1420 |