Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014252.3 | NP_055067.1 | 5, 6 | 954 | ||
GenBank mRNA | AF112968.1 | - | 5, 6 | 791 | ||
AF177333.1 | - | 5, 6 | 756 | |||
AK296862.1 | - | 6, 7 | 606 | |||
AK313602.1 | - | 5, 6 | 836 | |||
BC002702.2 | - | 5, 6 | 868 | 3614008 |