Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 15 | 2054 | ||
NM_001008212.1 | NP_001008213.1 | 14 | 1985 | |||
NM_001008213.1 | NP_001008214.1 | 15 | 2039 | |||
NM_021980.4 | NP_068815.2 | 13 | 1833 | |||
GenBank mRNA | AF049614.1 | - | 3 | 370 | ||
AF061034.1 | - | 15 | 1927 | |||
AF420371.1 | - | 15 | 1911 | |||
AF420372.1 | - | 13 | 1690 | |||
AF420373.1 | - | 14 | 1842 | |||
AK055403.1 | - | 14 | 2274 | |||
AK122746.1 | - | 5 | 1726 | |||
AK303715.1 | - | 13 | 1851 | |||
AK316171.1 | - | 13 | 1873 | |||
BC013876.2 | - | 13 | 1746 | 3831267 | ||
BC032762.1 | - | 15 | 1941 | 3457195 | ||
KU178502.1 | - | 12 | 1601 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ650013.1 | - | 540 | Chr. - - [-] on Build |