Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015120.4 | NP_055935.4 | 10 | 7941 | ||
GenBank mRNA | AB002326.3 | - | 3 | 1630 | ||
AJ417593.2 | - | 10 | 7941 | |||
AL833495.1 | - | 3 | 2113 | |||
AX751581.1 | - | 9 | 7815 | |||
BC035025.2 | - | 3 | 629 | |||
BC050330.1 | - | 3 | 629 | |||
BX647706.1 | - | 3 | 1854 | |||
BX648632.1 | - | 3 | 3693 |