Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271849.3 | XP_005271906.1 | 15, 16 | 2676 |
Gene Symbol: BMS1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027293.2 | 3, 4 | 367 | |||
GenBank mRNA | AK090629.1 | - | 3, 4 | 307 | ||
BX641141.1 | - | 3, 4 | 298 | |||
DB021899.1 | - | 3, 4 | 368 |