Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277126.1 | NP_001264055.1 | 3 | 970 | ||
NM_001277129.1 | NP_001264058.1 | 3 | 970 | |||
NM_144687.3 | NP_653288.1 | 3 | 970 | |||
XM_011527479.1 | XP_011525781.1 | 3 | 970 | |||
XM_011527480.1 | XP_011525782.1 | 3 | 970 | |||
XM_011527482.1 | XP_011525784.1 | 3 | 970 | |||
XM_017027460.1 | XP_016882949.1 | 3 | 970 | |||
XM_017027461.1 | XP_016882950.1 | 3 | 970 | |||
XM_017027462.1 | XP_016882951.1 | 3 | 970 | |||
XM_017027463.1 | XP_016882952.1 | 4 | 743 | |||
XM_017027464.1 | XP_016882953.1 | 3 | 541 | |||
XM_017027465.1 | XP_016882954.1 | 4 | 751 | |||
XM_017027466.1 | XP_016882955.1 | 4 | 748 | |||
XM_017027467.1 | XP_016882956.1 | 3 | 543 | |||
GenBank mRNA | AK095460.1 | - | 3 | 885 | ||
AK290772.1 | - | 3 | 752 | |||
AY095146.1 | - | 3 | 917 | |||
AY116204.1 | - | 3 | 961 | |||
AY116205.1 | - | 3 | 961 | |||
AY116206.1 | - | 3 | 961 | |||
AY116207.1 | - | 3 | 961 | |||
AY154467.1 | - | 3 | 917 | |||
BC028069.1 | - | 3 | 860 | 5212737 |