Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOLH1 SIRNA Assays ›
Gene Symbol:
FOLH1
Gene Name:
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases:
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
Chromosome Location:
Chr.11: 49145094 - 49208706 on Build GRCh38
UniGene ID:
Hs.645352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5327
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOLH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014986.1
NP_001014986.1
4, 5
783
NM_001193471.1
NP_001180400.1
5, 6
880
NM_001193472.1
NP_001180401.1
5, 6
880
NM_001193473.1
NP_001180402.1
3, 4
596
NM_004476.1
NP_004467.1
4, 5
783
XM_011519958.2
XP_011518260.2
5, 6
917
XM_017017432.1
XP_016872921.1
5, 6
915
XM_017017433.1
XP_016872922.1
5, 6
915
XM_017017434.1
XP_016872923.1
5, 6
581
XM_017017435.1
XP_016872924.1
5, 6
580
XM_017017436.1
XP_016872925.1
4, 5
815
XM_017017437.1
XP_016872926.1
4, 5
542
XM_017017438.1
XP_016872927.1
4, 5
815
XM_017017439.1
XP_016872928.1
5, 6
616
XM_017017440.1
XP_016872929.1
4, 5
486
XM_017017441.1
XP_016872930.1
4, 5
427
XM_017017442.1
XP_016872931.1
4, 5
553
XM_017017443.1
XP_016872932.1
4, 5
437
XM_017017444.1
XP_016872933.1
4, 5
717
XM_017017445.1
XP_016872934.1
5, 6
844
XM_017017446.1
XP_016872935.1
4, 5
815
XM_017017447.1
XP_016872936.1
4, 5
485
XM_017017448.1
XP_016872937.1
4, 5
477
XM_017017449.1
XP_016872938.1
4, 5
717
XM_017017450.1
XP_016872939.1
5, 6
844
XM_017017451.1
XP_016872940.1
4, 5
486
GenBank mRNA
AF176574.1
-
4, 5
660
AF254357.1
-
1, 2
45
AK295368.1
-
3, 4
528
AK295470.1
-
5, 6
812
AK300532.1
-
4, 5
715
AK312366.1
-
4, 5
711
AY101595.1
-
4, 5
522
BC025672.1
-
4, 5
681
5202715
BC108719.1
-
2, 3
287
DQ088979.1
-
4, 5
645
EF488811.2
-
5, 6
582
EF488812.1
-
5, 6
624
M99487.1
-
4, 5
783
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
folic acid-containing compound metabolic process
cellular amino acid biosynthetic process
C-terminal protein deglutamylation
metallocarboxypeptidase activity
peptidase activity
dipeptidase activity
metal ion binding
Ac-Asp-Glu binding
tetrahydrofolyl-poly(glutamate) polymer binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline