Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193333.2 | NP_001180262.1 | 10, 11 | 1500 | ||
NM_007074.3 | NP_009005.1 | 9, 10 | 1382 | |||
XM_011545714.2 | XP_011544016.1 | 9, 10 | 1195 | |||
XM_017022885.1 | XP_016878374.1 | 9, 10 | 1370 | |||
XM_017022886.1 | XP_016878375.1 | 9, 10 | 1192 | |||
GenBank mRNA | AB209221.1 | - | 3, 4 | 3766 | ||
AF495470.1 | - | 9, 10 | 1187 | |||
AK096332.1 | - | 8, 9 | 2377 | |||
AK314714.1 | - | 10, 11 | 1301 | |||
BC110374.1 | - | 9, 10 | 1199 | 5733853 | ||
BC126385.1 | - | 9, 10 | 1086 | 8992101 | ||
BC126387.1 | - | 9, 10 | 1086 | 8992103 | ||
D44497.1 | - | 9, 10 | 1165 | |||
U34690.1 | - | 9, 10 | 1157 | |||
X89109.1 | - | 9, 10 | 1158 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC064330.1 | - | 2842 | Chr. - - [-] on Build |