Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014714.3 | NP_055529.2 | 22 | 3206 | ||
XM_006720990.3 | XP_006721053.1 | 21 | 3124 | |||
XM_006720991.3 | XP_006721054.1 | 21 | 3314 | |||
XM_006720992.3 | XP_006721055.1 | 4 | 1680 | |||
XM_011522766.2 | XP_011521068.1 | 19 | 2662 | |||
XM_011522767.2 | XP_011521069.1 | 14 | 1926 | |||
XM_017023910.1 | XP_016879399.1 | 21 | 2897 | |||
XM_017023911.1 | XP_016879400.1 | 19 | 2757 | |||
GenBank mRNA | AB011162.1 | - | 21 | 2903 | ||
AK055133.1 | - | 4 | 538 | |||
AK299115.1 | - | 16 | 1966 | |||
AL080069.1 | - | 4 | 929 | |||
BC035577.1 | - | 20 | 2891 | 5552740 |