Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080.3 | NP_001071.1 | 8 | 1251 | ||
NM_170740.1 | NP_733936.1 | 9 | 1290 | |||
GenBank mRNA | AJ427354.2 | - | 9 | 1058 | ||
AJ427355.2 | - | 8 | 1223 | |||
AK290800.1 | - | 8 | 1369 | |||
AK294699.1 | - | 9 | 1028 | |||
AK314357.1 | - | 8 | 1251 | |||
AK315380.1 | - | 8 | 1346 | |||
BC034321.1 | - | 8 | 1251 | 4829533 | ||
KJ534766.1 | - | 8 | 1166 | |||
KJ534767.1 | - | 8 | 1223 | |||
KJ534768.1 | - | 8 | 1223 | |||
KJ534769.1 | - | 9 | 1262 | |||
L34820.1 | - | 5 | 587 | |||
Y11192.1 | - | 8 | 1223 |