Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015384.4 | NP_056199.2 | 24 | 5394 | ||
NM_133433.3 | NP_597677.2 | 24 | 5394 | |||
XM_005248280.3 | XP_005248337.1 | 24 | 5383 | |||
XM_005248282.4 | XP_005248339.3 | 18 | 4885 | |||
XM_006714467.2 | XP_006714530.1 | 24 | 5383 | |||
XM_006714468.2 | XP_006714531.1 | 23 | 5185 | |||
XM_011514015.1 | XP_011512317.1 | 24 | 5383 | |||
XM_017009329.1 | XP_016864818.1 | 24 | 5383 | |||
XM_017009330.1 | XP_016864819.1 | 16 | 7657 | |||
XM_017009331.1 | XP_016864820.1 | 23 | 3757 | |||
GenBank mRNA | AJ627032.1 | - | 24 | 5363 | ||
AJ640137.1 | - | 24 | 5363 | |||
KJ807791.1 | - | 10 | 1181 |