Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 20 | 2873 | ||
NM_001321185.1 | NP_001308114.1 | 20 | 2826 | |||
NM_007267.7 | NP_009198.4 | 20 | 2777 | |||
XM_005256995.1 | XP_005257052.1 | 20 | 2902 | |||
XM_011524255.1 | XP_011522557.1 | 19 | 2804 | |||
XM_011524256.1 | XP_011522558.1 | 19 | 2678 | |||
XM_011524257.2 | XP_011522559.1 | 14 | 1922 | |||
XM_011524258.1 | XP_011522560.1 | 11 | 1754 | |||
XM_017024107.1 | XP_016879596.1 | 19 | 2671 | |||
XM_017024108.1 | XP_016879597.1 | 19 | 2624 | |||
XM_017024109.1 | XP_016879598.1 | 18 | 2593 | |||
GenBank mRNA | AB002405.2 | - | 12 | 1585 | ||
AK074065.1 | - | 20 | 2776 | |||
AY099356.1 | - | 21 | 2775 | |||
AY099357.1 | - | 13 | 1722 | |||
BC018346.1 | - | 13 | 1741 | |||
BC023597.2 | - | 20 | 2757 | 4566166 | ||
BC035648.1 | - | 13 | 1797 | 5575949 |