Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178054.1 | NP_001171525.1 | 6 | 713 | ||
NM_013251.3 | NP_037383.1 | 7 | 767 | |||
NR_033654.1 | 8 | 884 | ||||
NR_135164.1 | 9 | 1042 | ||||
NR_135165.1 | 8 | 925 | ||||
NR_135166.1 | 7 | 871 | ||||
GenBank mRNA | AF186112.1 | - | 7 | 710 | ||
AF216586.1 | - | 7 | 697 | |||
AF537113.1 | - | 10 | 1179 | |||
AF537114.1 | - | 9 | 1002 | |||
AF537115.1 | - | 8 | 844 | |||
AF537116.1 | - | 9 | 1062 | |||
AF537117.1 | - | 8 | 885 | |||
AF537118.1 | - | 7 | 727 | |||
AF537119.1 | - | 8 | 1008 | |||
AF537120.1 | - | 7 | 831 | |||
AF537121.1 | - | 6 | 673 | |||
AX147032.1 | - | 7 | 613 | |||
AY358679.1 | - | 8 | 862 | |||
BC032145.2 | - | 7 | 748 | 4897300 |