Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282161.1 | NP_001269090.1 | 4, 5 | 489 | ||
NM_003336.3 | NP_003327.2 | 4, 5 | 496 | |||
NM_181762.2 | NP_861427.1 | 3, 4 | 406 | |||
GenBank mRNA | AK223045.1 | - | 4, 5 | 398 | ||
AK223064.1 | - | 4, 5 | 395 | |||
AK297696.1 | - | 3, 4 | 308 | |||
AK301675.1 | - | 3, 4 | 407 | |||
AK313092.1 | - | 4, 5 | 400 | |||
AL701289.1 | - | 4, 5 | 428 | |||
BC010175.2 | - | 4, 5 | 389 | 3051041 | ||
BC042021.1 | - | 1, 2 | 1273 | |||
BI093289.1 | - | 3, 4 | 196 | |||
DA068506.1 | - | 4, 5 | 393 | |||
DA599154.1 | - | 4, 5 | 496 | |||
M74524.1 | - | 4, 5 | 371 |