Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005522.4 | NP_005513.1 | 1 | 711 | ||
NM_153620.2 | NP_705873.2 | 2 | 508 | |||
GenBank mRNA | AK308067.1 | - | 2 | 456 | ||
AK313514.1 | - | 1 | 702 | |||
BC032547.2 | - | 1 | 679 | 5537563 | ||
S79869.1 | - | 1 | 611 | |||
S79871.1 | - | 2 | 408 | |||
S79910.1 | - | 1 | 611 | |||
U10421.1 | - | 1 | 617 | |||
U37431.1 | - | 1 | 611 |