Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133468.4 | NP_597725.1 | 15 | 2167 | ||
XM_005249633.2 | XP_005249690.1 | 13 | 1577 | |||
XM_017011800.1 | XP_016867289.1 | 13 | 1727 | |||
XM_017011801.1 | XP_016867290.1 | 12 | 1397 | |||
GenBank mRNA | AB075845.1 | - | 11 | 1434 | ||
AK289963.1 | - | 14 | 1907 | |||
AY324650.1 | - | 15 | 2166 | |||
BC060868.1 | - | 14 | 1844 | 30345290 | ||
BX380517.2 | - | 2 | 134 |