Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172810.1 | NP_001166281.1 | 14 | 2195 | ||
NM_023036.4 | NP_075462.3 | 14 | 2234 | |||
XM_011525125.2 | XP_011523427.1 | 15 | 2358 | |||
GenBank mRNA | AJ295276.1 | - | 13 | 2430 | ||
AL137526.1 | - | 6 | 1177 | |||
BC033013.1 | - | 13 | 1953 | 4826432 | ||
BC039582.1 | - | 14 | 2185 | 5752153 | ||
DB514781.1 | - | 2 | 374 |