Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127255.1 | NP_001120727.1 | 11 | 3135 | ||
NM_139176.3 | NP_631915.2 | 11 | 3051 | |||
NM_206828.3 | NP_996611.2 | 10 | 2964 | |||
XM_006723075.3 | XP_006723138.1 | 12 | 3370 | |||
XM_006723076.3 | XP_006723139.1 | 11 | 3333 | |||
XM_011526596.2 | XP_011524898.1 | 13 | 3577 | |||
XM_011526599.2 | XP_011524901.1 | 13 | 3588 | |||
XM_011526601.2 | XP_011524903.1 | 12 | 3406 | |||
GenBank mRNA | AF464765.1 | - | 9 | 2934 | ||
AK298630.1 | - | 9 | 2205 | |||
AK302655.1 | - | 13 | 3546 | |||
AK313964.1 | - | 13 | 3530 | |||
AY154462.1 | - | 10 | 2958 | |||
BC109124.2 | - | 11 | 3090 | 40036025 | ||
BC109125.1 | - | 11 | 3127 | 40036028 |