Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000547.5 | NP_000538.3 | 4, 5 | 443 | ||
NM_001206744.1 | NP_001193673.1 | 4, 5 | 438 | |||
NM_001206745.1 | NP_001193674.1 | 4, 5 | 438 | |||
NM_175719.3 | NP_783650.1 | 4, 5 | 443 | |||
NM_175721.3 | NP_783652.1 | 3, 4 | 353 | |||
NM_175722.3 | NP_783653.1 | 3, 4 | 353 | |||
XM_011510379.2 | XP_011508681.1 | 3, 4 | 353 | |||
XM_011510380.2 | XP_011508682.1 | 3, 4 | 353 | |||
XM_011510381.2 | XP_011508683.1 | 3, 4 | 353 | |||
GenBank mRNA | AB208960.1 | - | 1, 2 | 139 | ||
AF439430.1 | - | 3, 4 | 352 | |||
AF533528.1 | - | 2, 3 | 271 | |||
BC063107.1 | - | 4, 5 | 445 | |||
BC095448.1 | - | 4, 5 | 436 | 30329791 | ||
DA958481.1 | - | 3, 4 | 379 | |||
J02969.1 | - | 4, 5 | 424 | |||
J02970.1 | - | 4, 5 | 424 | |||
M17755.2 | - | 4, 5 | 436 | |||
X17358.1 | - | 4, 5 | 417 | |||
Y00406.1 | - | 4, 5 | 392 |