Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIKFYVE SIRNA Assays ›
Gene Symbol:
PIKFYVE
Gene Name:
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases:
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location:
Chr.2: 208266178 - 208358751 on Build GRCh38
UniGene ID:
Hs.744997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s532910
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIKFYVE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178000.1
NP_001171471.1
6
807
NM_015040.3
NP_055855.2
6
807
NM_152671.3
NP_689884.1
4
516
XM_011510778.2
XP_011509080.1
7
857
XM_011510779.2
XP_011509081.1
7
790
XM_011510780.2
XP_011509082.1
7
875
XM_011510781.2
XP_011509083.1
7
816
XM_011510782.2
XP_011509084.1
7
857
XM_011510783.2
XP_011509085.1
7
857
XM_011510784.2
XP_011509086.1
7
875
XM_011510785.2
XP_011509087.1
7
815
XM_011510786.2
XP_011509088.1
5
524
XM_011510787.1
XP_011509089.1
5
533
XM_011510788.1
XP_011509090.1
4
516
XM_011510789.2
XP_011509091.1
3
338
XM_011510790.1
XP_011509092.1
5
635
XM_011510792.2
XP_011509094.1
7
857
XM_017003568.1
XP_016859057.1
6
711
XM_017003569.1
XP_016859058.1
6
711
XM_017003570.1
XP_016859059.1
4
542
XM_017003571.1
XP_016859060.1
3
338
XM_017003572.1
XP_016859061.1
5
511
GenBank mRNA
AK127769.1
-
6
711
AY457063.1
-
6
811
BC017736.2
-
5
635
BC032389.1
-
4
516
BC125052.1
-
5
538
BC125053.1
-
6
796
EU794609.1
-
6
811
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AF102777, AK173077, AK080527, AK165350, AK139116, and others.
Panther Classification:
Molecular Function -
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
receptor-mediated endocytosis
myelin assembly
protein localization to nucleus
intracellular signal transduction
phosphatidylinositol-3-phosphate biosynthetic process
retrograde transport, endosome to Golgi
phosphatidylinositol phosphorylation
phosphatidylinositol 5-phosphate metabolic process
regulation of autophagosome assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
zinc ion binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline