Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012434.4 | NP_036566.1 | 8 | 1145 | ||
XM_005248710.3 | XP_005248767.1 | 8 | 1167 | |||
XM_005248711.2 | XP_005248768.1 | 8 | 1399 | |||
XM_011535750.2 | XP_011534052.1 | 8 | 1103 | |||
XM_017010730.1 | XP_016866219.1 | 7 | 1311 | |||
XM_017010731.1 | XP_016866220.1 | 7 | 1161 | |||
GenBank mRNA | AF244577.1 | - | 9 | 1402 | ||
AJ387747.1 | - | 8 | 1287 | |||
AK025880.1 | - | 2 | 58 | |||
AK075320.1 | - | 9 | 1201 | |||
AK294068.1 | - | 7 | 1016 | |||
AK315789.1 | - | 8 | 1103 | |||
BC020961.2 | - | 8 | 1104 | 3847279 |