Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000132.3 | NP_000123.1 | 25 | 6975 | ||
NM_019863.2 | NP_063916.1 | 4 | 544 | |||
GenBank mRNA | AK289947.1 | - | 4 | 474 | ||
AK313707.1 | - | 4 | 429 | |||
AX746360.1 | - | 25 | 6804 | |||
BC022513.1 | - | 4 | 463 | |||
BC064380.1 | - | 4 | 523 | |||
BC098389.1 | - | 4 | 542 | |||
BC111967.1 | - | 4 | 432 | |||
BC111969.1 | - | 4 | 431 | |||
K01740.1 | - | 25 | 6954 | |||
KT878539.1 | - | 2 | 225 | |||
M14113.1 | - | 25 | 6975 | |||
M90707.1 | - | 4 | 439 | |||
X01179.1 | - | 25 | 6913 |