Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016350.4 | NP_057434.4 | 5 | 484 | ||
NM_020921.3 | NP_065972.3 | 5 | 484 | |||
NM_182944.2 | NP_891989.2 | 5 | 484 | |||
NM_182946.1 | NP_891991.1 | 5 | 484 | |||
XM_011536817.1 | XP_011535119.1 | 5 | 458 | |||
XM_011536819.2 | XP_011535121.1 | 5 | 458 | |||
XM_011536822.1 | XP_011535124.1 | 5 | 458 | |||
XM_011536823.2 | XP_011535125.1 | 4 | 335 | |||
GenBank mRNA | AB046785.2 | - | 5 | 409 | ||
AF212162.2 | - | 4 | 513 | |||
AF223937.1 | - | 5 | 491 | |||
AF223938.1 | - | 5 | 491 | |||
AF223939.1 | - | 5 | 491 | |||
AF302773.2 | - | 5 | 484 | |||
AY736182.1 | - | 5 | 484 | |||
BC065521.1 | - | 5 | 394 | |||
BC090932.1 | - | 3 | 328 | 3090109 |