Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 11 | 1202 | ||
NR_024265.1 | 11 | 1345 | ||||
XM_006713705.3 | XP_006713768.1 | 11 | 1154 | |||
XM_017006878.1 | XP_016862367.1 | 11 | 1317 | |||
XM_017006879.1 | XP_016862368.1 | 12 | 1463 | |||
XM_017006880.1 | XP_016862369.1 | 12 | 1320 | |||
GenBank mRNA | AK027012.1 | - | 2 | 131 | ||
AK075444.1 | - | 11 | 1138 | |||
AK126736.1 | - | 3 | 2831 | |||
AK295984.1 | - | 11 | 1320 | |||
AK315367.1 | - | 11 | 1177 | |||
AY298903.1 | - | 11 | 1188 | |||
AY358581.1 | - | 11 | 1145 | |||
BC030614.2 | - | 11 | 1188 | 4796951 | ||
BC048810.1 | - | 11 | 1146 | 5243487 |