Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 11 | 3350 | ||
NR_024265.1 | 11 | 3493 | ||||
XM_006713705.3 | XP_006713768.1 | 11 | 3302 | |||
XM_017006878.1 | XP_016862367.1 | 11 | 3465 | |||
XM_017006879.1 | XP_016862368.1 | 12 | 3611 | |||
XM_017006880.1 | XP_016862369.1 | 12 | 3468 | |||
GenBank mRNA | AK027012.1 | - | 2 | 2276 | ||
AY298903.1 | - | 11 | 3336 |