Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312686.1 | NP_001299615.1 | 16 | 2466 | ||
NM_014813.2 | NP_055628.1 | 15 | 2354 | |||
XM_005271369.2 | XP_005271426.1 | 14 | 2033 | |||
XM_017002952.1 | XP_016858441.1 | 15 | 2005 |