Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110556.1 | NP_001104026.1 | 29, 30 | 5200 | ||
XM_011531127.1 | XP_011529429.1 | 29, 30 | 5195 | |||
XM_011531129.1 | XP_011529431.1 | 29, 30 | 5195 | |||
XM_011531131.1 | XP_011529433.1 | 28, 29 | 4994 | |||
GenBank mRNA | AB191260.1 | - | 28, 29 | 4928 | ||
AB371574.1 | - | 28, 29 | 4928 | |||
AB371576.1 | - | 28, 29 | 4928 | |||
AB371577.1 | - | 28, 29 | 4923 | |||
AB593010.1 | - | 28, 29 | 5063 | |||
AK300165.1 | - | 2, 3 | 236 | |||
GU727643.1 | - | 29, 30 | 5200 | |||
X53416.1 | - | 29, 30 | 5123 |