Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145320.1 | NP_001138792.1 | 8 | 1284 | ||
NM_014694.3 | NP_055509.2 | 8 | 942 | |||
XM_005272237.3 | XP_005272294.1 | 8 | 1139 | |||
XM_005272238.3 | XP_005272295.1 | 7 | 781 | |||
XM_005272239.2 | XP_005272296.1 | 8 | 817 | |||
XM_011519241.2 | XP_011517543.2 | 7 | 960 | |||
XM_011519242.2 | XP_011517544.1 | 7 | 816 | |||
GenBank mRNA | AB011177.1 | - | 8 | 1284 | ||
BC050544.1 | - | 8 | 942 | 6196393 |