Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 10, 11 | 1734 | ||
XM_005263565.4 | XP_005263622.1 | 10, 11 | 1869 | |||
XM_005263568.4 | XP_005263625.1 | 10, 11 | 1869 | |||
XM_011510490.2 | XP_011508792.1 | 10, 11 | 1769 | |||
XM_017003164.1 | XP_016858653.1 | 10, 11 | 1739 | |||
XM_017003165.1 | XP_016858654.1 | 11, 12 | 1838 | |||
GenBank mRNA | AK294196.1 | - | 2, 3 | 180 | ||
AK308695.1 | - | 2, 3 | 182 | |||
AK316584.1 | - | 10, 11 | 1741 | |||
BC114917.1 | - | 10, 11 | 1448 | 40068588 | ||
BC114918.1 | - | 10, 11 | 1448 | 40012335 | ||
U08023.1 | - | 10, 11 | 1749 |