Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022167.3 | NP_071450.2 | 10 | 2373 | ||
NR_110010.1 | 9 | 2177 | ||||
XM_005257572.4 | XP_005257629.1 | 10 | 2687 | |||
GenBank mRNA | AJ277442.1 | - | 10 | 2373 | ||
AY358090.1 | - | 9 | 2073 | |||
BC052262.1 | - | 10 | 2293 | 6380013 |