Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533054
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
19
4977
NM_004801.4
NP_004792.1
18
4857
NM_138735.2
NP_620072.1
2
1082
XM_005264642.3
XP_005264699.1
18
4878
XM_005264643.3
XP_005264700.1
17
4833
XM_006712137.3
XP_006712200.1
18
4878
XM_006712140.3
XP_006712203.2
19
4905
XM_011533167.2
XP_011531469.1
18
4878
XM_011533171.2
XP_011531473.1
18
4857
XM_011533172.2
XP_011531474.1
17
4851
XM_011533174.2
XP_011531476.1
17
4833
XM_011533175.2
XP_011531477.1
16
4821
XM_011533177.2
XP_011531479.1
18
4878
XM_011533178.2
XP_011531480.1
15
4788
XM_011533180.2
XP_011531482.1
18
4878
XM_011533183.1
XP_011531485.1
12
2560
XM_017005303.1
XP_016860792.1
19
4905
XM_017005304.1
XP_016860793.1
19
4902
XM_017005305.1
XP_016860794.1
19
4905
XM_017005306.1
XP_016860795.1
18
4893
XM_017005307.1
XP_016860796.1
18
4887
XM_017005308.1
XP_016860797.1
19
4884
XM_017005309.1
XP_016860798.1
18
4878
XM_017005310.1
XP_016860799.1
17
4875
XM_017005311.1
XP_016860800.1
18
4860
XM_017005312.1
XP_016860801.1
18
4854
XM_017005313.1
XP_016860802.1
18
4857
XM_017005314.1
XP_016860803.1
16
4845
XM_017005315.1
XP_016860804.1
17
4851
XM_017005316.1
XP_016860805.1
17
4842
XM_017005317.1
XP_016860806.1
17
4845
XM_017005318.1
XP_016860807.1
17
4833
XM_017005319.1
XP_016860808.1
17
4830
XM_017005320.1
XP_016860809.1
17
4830
XM_017005321.1
XP_016860810.1
19
4905
XM_017005322.1
XP_016860811.1
19
4905
XM_017005323.1
XP_016860812.1
14
4791
XM_017005324.1
XP_016860813.1
17
4851
XM_017005325.1
XP_016860814.1
17
4851
XM_017005326.1
XP_016860815.1
17
4839
XM_017005327.1
XP_016860816.1
17
4833
XM_017005328.1
XP_016860817.1
16
4806
XM_017005329.1
XP_016860818.1
19
4905
XM_017005330.1
XP_016860819.1
14
4791
XM_017005331.1
XP_016860820.1
14
4776
XM_017005332.1
XP_016860821.1
13
4746
XM_017005333.1
XP_016860822.1
13
4746
XM_017005334.1
XP_016860823.1
15
2726
XM_017005335.1
XP_016860824.1
2
6072
XM_017005336.1
XP_016860825.1
2
6073
XM_017005337.1
XP_016860826.1
2
6072
GenBank mRNA
AB011150.2
-
16
3539
AB035356.1
-
17
3572
AF064842.1
-
2
1081
AK093260.1
-
12
2560
BC125179.1
-
3
138
BC125180.1
-
3
501
BC150247.1
-
16
3560
9007472
EF539882.1
-
19
4840
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline