Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042355.1 | NP_001035814.1 | 8, 9 | 1785 | ||
NM_001042412.1 | NP_001035877.1 | 8, 9 | 1809 | |||
NM_015247.2 | NP_056062.1 | 10, 11 | 1931 | |||
XM_005255812.2 | XP_005255869.1 | 9, 10 | 1882 | |||
XM_006721149.1 | XP_006721212.1 | 9, 10 | 1946 | |||
XM_011522907.2 | XP_011521209.1 | 8, 9 | 1708 | |||
XM_011522908.1 | XP_011521210.1 | 8, 9 | 1770 | |||
XM_011522911.2 | XP_011521213.1 | 8, 9 | 1800 | |||
XM_017022977.1 | XP_016878466.1 | 8, 9 | 2080 | |||
XM_017022978.1 | XP_016878467.1 | 8, 9 | 1876 | |||
XM_017022979.1 | XP_016878468.1 | 8, 9 | 1836 | |||
XM_017022980.1 | XP_016878469.1 | 8, 9 | 1869 | |||
XM_017022981.1 | XP_016878470.1 | 8, 9 | 1737 | |||
GenBank mRNA | AF161542.1 | - | 7, 8 | 1757 | ||
AJ250014.1 | - | 10, 11 | 1907 | |||
AK000187.1 | - | 3, 4 | 587 | |||
AK024212.1 | - | 6, 7 | 1243 | |||
AK024348.1 | - | 6, 7 | 1292 | |||
AK056226.1 | - | 8, 9 | 1740 | |||
AK225202.1 | - | 9, 10 | 1902 | |||
AK292975.1 | - | 8, 9 | 1767 | |||
AK310832.1 | - | 8, 9 | 1757 | |||
BC012342.1 | - | 8, 9 | 1783 | 4552767 |