Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000431.3 | NP_000422.1 | 11 | 1291 | ||
NM_001114185.2 | NP_001107657.1 | 11 | 1282 | |||
NM_001301182.1 | NP_001288111.1 | 10 | 1135 | |||
XM_011538372.2 | XP_011536674.2 | 11 | 1399 | |||
XM_017019313.1 | XP_016874802.1 | 10 | 1088 | |||
XM_017019314.1 | XP_016874803.1 | 11 | 1757 | |||
GenBank mRNA | AB209722.1 | - | 11 | 1174 | ||
AF217536.1 | - | 10 | 1114 | |||
AK023087.1 | - | 4 | 1627 | |||
AK293130.1 | - | 9 | 858 | |||
AK295338.1 | - | 11 | 993 | |||
AK315678.1 | - | 11 | 1170 | |||
BC016140.1 | - | 11 | 1134 | 3921543 | ||
KU178205.1 | - | 10 | 1098 | |||
KU178206.1 | - | 9 | 942 | |||
M88468.1 | - | 11 | 1189 |