Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014080.4 | NP_054799.4 | 20 | 2793 | ||
XM_005254421.2 | XP_005254478.1 | 20 | 2793 | |||
GenBank mRNA | AB209010.1 | - | 14 | 4321 | ||
AF181972.1 | - | 12 | 1576 | |||
AF230496.1 | - | 20 | 2775 | |||
AF267981.1 | - | 20 | 2793 |