Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015506.2 | NP_056321.2 | 4 | 901 | ||
XM_005270724.4 | XP_005270781.1 | 3 | 660 | |||
XM_011541204.2 | XP_011539506.1 | 4 | 810 | |||
GenBank mRNA | AL080062.1 | - | 4 | 772 | ||
BC006122.1 | - | 4 | 744 | 3535453 |