Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039141.2 | NP_001034230.1 | 13 | 5725 | ||
NM_007032.5 | NP_008963.3 | 3 | 319 | |||
NM_138632.2 | NP_619538.2 | 3 | 319 | |||
GenBank mRNA | AF281030.1 | - | 3 | 303 | ||
BC003618.2 | - | 3 | 241 | |||
BC022200.1 | - | 3 | 313 | |||
BQ880866.1 | - | 319 | ||||
BX342673.2 | - | 3 | 314 | |||
DQ228003.1 | - | 11 | 5388 | |||
DQ228005.1 | - | 12 | 5590 |