Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000378.4 | NP_000369.3 | 5 | 1218 | ||
NM_001198551.1 | NP_001185480.1 | 6 | 721 | |||
NM_001198552.1 | NP_001185481.1 | 5 | 670 | |||
NM_024424.3 | NP_077742.2 | 6 | 1269 | |||
NM_024426.4 | NP_077744.3 | 6 | 1269 | |||
GenBank mRNA | AB971668.1 | - | 6 | 1428 | ||
AK093168.1 | - | 5 | 666 | |||
AK291736.1 | - | 6 | 1269 | |||
BC032861.2 | - | 6 | 716 | 4827849 | ||
BC046461.1 | - | 6 | 1274 | |||
BG718348.1 | - | 6 | 726 | |||
M30393.1 | - | 5 | 572 | |||
X51630.1 | - | 6 | 1255 |