Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173466.1 | NP_001166937.1 | 3, 4 | 478 | ||
NM_015665.5 | NP_056480.1 | 3, 4 | 478 | |||
XM_011538778.1 | XP_011537080.1 | 3, 4 | 410 | |||
XM_011538780.1 | XP_011537082.1 | 3, 4 | 410 | |||
GenBank mRNA | AF226048.1 | - | 3, 4 | 397 | ||
AJ289857.1 | - | 3, 4 | 405 | |||
AK000833.1 | - | 3, 4 | 405 | |||
AK094302.1 | - | 2, 3 | 757 | |||
AK222509.1 | - | 3, 4 | 405 | |||
AK293342.1 | - | 3, 4 | 435 | |||
AY237818.1 | - | 3, 4 | 446 | |||
BC000659.2 | - | 3, 4 | 440 | 3347423 | ||
BC018836.2 | - | 3, 4 | 440 | |||
BT006912.1 | - | 3, 4 | 315 | |||
DC393722.1 | - | 3, 4 | 478 |