Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 7 | 691 | ||
NM_001142618.1 | NP_001136090.1 | 7 | 673 | |||
NM_001142619.1 | NP_001136091.1 | 7 | 664 | |||
NM_001142620.1 | NP_001136092.1 | 6 | 874 | |||
NM_001199040.1 | NP_001185969.1 | 7 | 807 | |||
NM_001199041.1 | NP_001185970.1 | 7 | 818 | |||
NM_001199042.1 | NP_001185971.1 | 7 | 922 | |||
NM_022369.3 | NP_071764.3 | 7 | 627 | |||
XM_011521883.1 | XP_011520185.1 | 7 | 593 | |||
XM_011521884.1 | XP_011520186.1 | 7 | 734 | |||
XM_011521885.2 | XP_011520187.1 | 7 | 568 | |||
XM_017022478.1 | XP_016877967.1 | 7 | 542 | |||
XM_017022479.1 | XP_016877968.1 | 7 | 633 | |||
XM_017022480.1 | XP_016877969.1 | 7 | 657 | |||
GenBank mRNA | AF352728.1 | - | 7 | 493 | ||
AF352729.1 | - | 7 | 603 | |||
AF370419.2 | - | 7 | 599 | |||
AK022603.1 | - | 3 | 177 | |||
AK056125.1 | - | 7 | 688 | |||
AK091152.1 | - | 7 | 717 | |||
AK291966.1 | - | 7 | 563 | |||
AK298921.1 | - | 7 | 562 | |||
AK298928.1 | - | 5 | 231 | |||
AK299191.1 | - | 7 | 807 | |||
AK302932.1 | - | 7 | 818 | |||
AK309676.1 | - | 7 | 902 | |||
AK310878.1 | - | 7 | 550 | |||
AY358748.1 | - | 7 | 603 | |||
AY359089.1 | - | 7 | 493 | |||
BC015881.2 | - | 6 | 822 | |||
BC025256.1 | - | 7 | 599 | 4892821 | ||
BX537413.1 | - | 7 | 569 |