Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 13 | 1365 | ||
NM_001142618.1 | NP_001136090.1 | 13 | 1347 | |||
NM_001142619.1 | NP_001136091.1 | 13 | 1338 | |||
NM_001199040.1 | NP_001185969.1 | 13 | 1481 | |||
NM_001199041.1 | NP_001185970.1 | 13 | 1492 | |||
NM_001199042.1 | NP_001185971.1 | 13 | 1596 | |||
NM_022369.3 | NP_071764.3 | 13 | 1301 | |||
XM_011521883.1 | XP_011520185.1 | 13 | 1267 | |||
XM_011521884.1 | XP_011520186.1 | 13 | 1408 | |||
XM_017022478.1 | XP_016877967.1 | 13 | 1216 | |||
XM_017022479.1 | XP_016877968.1 | 13 | 1307 | |||
XM_017022480.1 | XP_016877969.1 | 13 | 1331 | |||
GenBank mRNA | AF352728.1 | - | 13 | 1167 | ||
AF352729.1 | - | 13 | 1277 | |||
AF370419.2 | - | 13 | 1273 | |||
AK022603.1 | - | 9 | 851 | |||
AK056125.1 | - | 13 | 1362 | |||
AK091152.1 | - | 13 | 1391 | |||
AK092227.1 | - | 8 | 828 | |||
AK291966.1 | - | 13 | 1237 | |||
AK298921.1 | - | 13 | 1236 | |||
AK298928.1 | - | 11 | 905 | |||
AK299191.1 | - | 13 | 1481 | |||
AK302932.1 | - | 13 | 1492 | |||
AK309676.1 | - | 13 | 1576 | |||
AK310878.1 | - | 12 | 1354 | |||
AY358748.1 | - | 13 | 1277 | |||
AY359089.1 | - | 13 | 1167 | |||
BC025256.1 | - | 13 | 1273 | 4892821 | ||
BX537413.1 | - | 13 | 1243 |