Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 10, 11 | 1119 | ||
NM_001142618.1 | NP_001136090.1 | 10, 11 | 1101 | |||
NM_001142619.1 | NP_001136091.1 | 10, 11 | 1092 | |||
NM_001199040.1 | NP_001185969.1 | 10, 11 | 1235 | |||
NM_001199041.1 | NP_001185970.1 | 10, 11 | 1246 | |||
NM_001199042.1 | NP_001185971.1 | 10, 11 | 1350 | |||
NM_022369.3 | NP_071764.3 | 10, 11 | 1055 | |||
XM_011521883.1 | XP_011520185.1 | 10, 11 | 1021 | |||
XM_011521884.1 | XP_011520186.1 | 10, 11 | 1162 | |||
XM_011521885.2 | XP_011520187.1 | 10, 11 | 996 | |||
XM_017022478.1 | XP_016877967.1 | 10, 11 | 970 | |||
XM_017022479.1 | XP_016877968.1 | 10, 11 | 1061 | |||
XM_017022480.1 | XP_016877969.1 | 10, 11 | 1085 | |||
GenBank mRNA | AF352728.1 | - | 10, 11 | 921 | ||
AF352729.1 | - | 10, 11 | 1031 | |||
AF370419.2 | - | 10, 11 | 1027 | |||
AK022603.1 | - | 6, 7 | 605 | |||
AK056125.1 | - | 10, 11 | 1116 | |||
AK091152.1 | - | 10, 11 | 1145 | |||
AK092227.1 | - | 5, 6 | 582 | |||
AK291966.1 | - | 10, 11 | 991 | |||
AK298921.1 | - | 10, 11 | 990 | |||
AK299191.1 | - | 10, 11 | 1235 | |||
AK302932.1 | - | 10, 11 | 1246 | |||
AK309676.1 | - | 10, 11 | 1330 | |||
AK310878.1 | - | 10 | 1108 | |||
AY358748.1 | - | 10, 11 | 1031 | |||
AY359089.1 | - | 10, 11 | 921 | |||
BC025256.1 | - | 10, 11 | 1027 | 4892821 | ||
BX537413.1 | - | 10, 11 | 997 |