Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 5 | 748 | ||
XM_006716392.2 | XP_006716455.1 | 5 | 931 | |||
GenBank mRNA | AF035277.1 | - | 5 | 596 | ||
AK090513.1 | - | 4 | 932 | |||
AK315122.1 | - | 5 | 618 | |||
BC010550.1 | - | 5 | 549 | 4153601 | ||
CR456803.1 | - | 5 | 484 | |||
CR541850.1 | - | 5 | 484 | |||
KU178337.1 | - | 5 | 484 | |||
S79669.1 | - | 5 | 636 | |||
U17280.1 | - | 5 | 610 |