Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077416.2 | NP_001070884.2 | 3 | 681 | ||
NM_001077418.2 | NP_001070886.1 | 4 | 584 | |||
NR_074083.1 | 4 | 707 | ||||
GenBank mRNA | AK025820.1 | - | 4 | 556 | ||
AK096650.1 | - | 3 | 633 | |||
AK290483.1 | - | 4 | 570 | |||
AK298994.1 | - | 1 | 278 | |||
AK307263.1 | - | 4 | 673 | |||
AY358612.1 | - | 4 | 580 | |||
BC010609.1 | - | 4 | 538 | 4214587 | ||
BC016401.1 | - | 3 | 216 | |||
BC063677.1 | - | 4 | 689 | |||
BU173659.1 | - | 4 | 648 |