Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 9 | 1300 | ||
NM_001030312.2 | NP_001025483.1 | 5 | 883 | |||
NM_001030313.2 | NP_001025484.1 | 6 | 1015 | |||
NM_001160277.1 | NP_001153749.1 | 8 | 1168 | |||
NM_201548.4 | NP_963842.1 | 8 | 1222 | |||
NR_027689.1 | 6 | 1026 | ||||
NR_027690.1 | 7 | 1158 | ||||
GenBank mRNA | AJ640141.1 | - | 8 | 1139 | ||
AJ697855.1 | - | 8 | 1121 | |||
AJ697858.1 | - | 6 | 941 | |||
AK129976.1 | - | 5 | 465 | |||
AK293844.1 | - | 8 | 1169 | |||
AY357073.1 | - | 8 | 1121 | |||
AY690329.1 | - | 9 | 1199 | |||
AY690330.1 | - | 5 | 782 | |||
AY690331.1 | - | 6 | 914 | |||
AY690332.1 | - | 6 | 925 | |||
AY690333.1 | - | 7 | 1057 | |||
BC137498.1 | - | 8 | 1148 | 9021498 | ||
BC137499.1 | - | 8 | 1148 | 9021499 | ||
HQ426666.1 | - | 8 | 1028 | |||
HQ426667.1 | - | 7 | 964 | |||
HQ426668.1 | - | 6 | 832 | |||
HQ426669.1 | - | 6 | 821 | |||
HQ426670.1 | - | 5 | 689 | |||
HQ426671.1 | - | 9 | 1028 | |||
HQ426672.1 | - | 8 | 950 | |||
HQ426673.1 | - | 7 | 886 | |||
HQ426674.1 | - | 6 | 754 | |||
HQ426675.1 | - | 6 | 743 | |||
HQ426676.1 | - | 5 | 611 | |||
HQ426677.1 | - | 6 | 558 | |||
HQ426678.1 | - | 9 | 1275 | |||
HQ426679.1 | - | 7 | 883 | |||
JF290420.1 | - | 8 | 1108 |